ENA053246385 Enamine

[(5-bromothiophen-2-yl)methyl](ethyl)amine

  • MDL number MFCD06385604
  • Cas Number : 851788-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12313
Availability details for
Quantity
  • Availability
  • Add to Cart