ENA053246386 Enamine

(methyl{[2-phenylvinyl]sulfonyl}amino)acetic acid

  • MDL number MFCD06655291
  • Cas Number : 851879-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12314
Availability details for
Quantity
  • Availability
  • Add to Cart