ENA053246389 Enamine

ethyl 3-amino-6-oxo-1H,6H,7H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD06660518
  • Cas Number : 851879-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12317
Availability details for
Quantity
  • Availability
  • Add to Cart