ENA053246391 Enamine

1-[(5-mercapto-4-pyridin-2-yl-4H-1,2,4-triazol-3-yl)methyl]pyrrolidin-2-one

  • MDL number MFCD06655292
  • Cas Number : 851879-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12319
Availability details for
Quantity
  • Availability
  • Add to Cart