ENA053246392 Enamine

2-(4-carbamoyl-1-oxo-1,2-dihydrophthalazin-2-yl)acetic acid

  • MDL number MFCD06655293
  • Cas Number : 851879-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12320
Availability details for
Quantity
  • Availability
  • Add to Cart