ENA053246394 Enamine

4,5-dimethyl-1,3-oxazol-2-amine

  • MDL number MFCD06655295
  • Cas Number : 45529-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12322

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-12322
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart