ENA053246407 Enamine

5-(dimethylsulfamoyl)-2-(piperidin-1-yl)benzoic acid

  • MDL number MFCD06655300
  • Cas Number : 77266-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12335
Availability details for
Quantity
  • Availability
  • Add to Cart