ENA053246412 Enamine

ethyl 4-(2-methyl-4-oxoquinazolin-3(4H)-yl)benzoate

  • MDL number MFCD01816208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12340
Availability details for
Quantity
  • Availability
  • Add to Cart