ENA053246415 Enamine

4-(4-oxo-3,4-dihydroquinazolin-3-yl)benzoic acid

  • MDL number MFCD03473182
  • Cas Number : 94242-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12343
Availability details for
Quantity
  • Availability
  • Add to Cart