ENA053246417 Enamine

N-(2,3-dimethylphenyl)-2-(piperazin-1-yl)acetamide

  • MDL number MFCD06655303
  • Cas Number : 851903-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12345
Availability details for
Quantity
  • Availability
  • Add to Cart