ENA053246419 Enamine

1-(1,1-dioxo-1lambda6-thiolan-3-yl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06655305
  • Cas Number : 955043-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12347
Availability details for
Quantity
  • Availability
  • Add to Cart