ENA053246422 Enamine

4-[(dimethyl-1,2-oxazol-4-yl)methoxy]benzaldehyde

  • MDL number MFCD06253775
  • Cas Number : 610260-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12350
Availability details for
Quantity
  • Availability
  • Add to Cart