ENA053246424 Enamine

2-(4-chloro-3-nitrophenyl)-1,3-dithiolane

  • MDL number MFCD06655308
  • Cas Number : 852218-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12352

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12352
Availability details for
Quantity
  • Availability
  • Add to Cart