ENA053246437 Enamine

1-[(1,3-benzothiazol-2-yl)methyl]cyclopentane-1-carboxylic acid

  • MDL number MFCD06655314
  • Cas Number : 852033-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12366
Availability details for
Quantity
  • Availability
  • Add to Cart