ENA053246438 Enamine

(4-oxo-6-phenylthieno[2,3-d]pyrimidin-3(4H)-yl)acetic acid

  • MDL number MFCD02730281
  • Cas Number : 371233-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12367
Availability details for
Quantity
  • Availability
  • Add to Cart