ENA053246453 Enamine

2-chloro-N-[4-chloro-3-(trifluoromethyl)phenyl]-2-phenylacetamide

  • MDL number MFCD03970398
  • Cas Number : 568553-09-3
  • Molecular Weight : 348.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-01988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01988
Availability details for
Quantity
  • Availability
  • Add to Cart