ENA053246456 Enamine

2-[2-(thiophene-2-amido)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD00832128
  • Cas Number : 329695-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12385
Availability details for
Quantity
  • Availability
  • Add to Cart