ENA053246457 Enamine

2-(2-benzamido-1,3-thiazol-4-yl)acetic acid

  • MDL number MFCD00832130
  • Cas Number : 329698-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12386
Availability details for
Quantity
  • Availability
  • Add to Cart