ENA053246460 Enamine

2-(6-bromohexyl)-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD00023098
  • Cas Number : 24566-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12389

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-12389
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart