ENA053246471 Enamine

4,5-dimethylthiophene-2-carboxylic acid

  • MDL number MFCD00512748
  • Cas Number : 40808-24-0
  • Molecular Weight : 156.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-211
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11978

Safety Information

signalword Warning
pcodes P264,P280,P305+P351+P338,P337+P313
MSDS https://www.enaminestore.com/catalog/EN300-11978
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart