ENA053246474 Enamine

4-[(furan-2-yl)methyl]-5-(2-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06380331
  • Cas Number : 851398-38-4
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11980
Availability details for
Quantity
  • Availability
  • Add to Cart