ENA053246481 Enamine

3-amino-4,5-dimethylbenzene-1-sulfonamide

  • MDL number MFCD07311123
  • Cas Number : 101251-33-6
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 159-160
Iupac Name 3-amino-4,5-dimethylbenzenesulfonamide
Inchi Code 1S/C8H12N2O2S/c1-5-3-7(13(10,11)12)4-8(9)6(5)2/h3-4H,9H2,1-2H3,(H2,10,11,12)
Inchi Key YXQMLWCILAYJQY-UHFFFAOYSA-N

Description

Melting Point 159-160
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12410

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-12410
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart