ENA053246486 Enamine

2-(4-methoxyphenyl)-4-nitrophenol

  • MDL number MFCD06380329
  • Cas Number : 63801-89-8
  • Molecular Weight : 245.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 83-84
Iupac Name 4'-methoxy-5-nitro[1,1'-biphenyl]-2-ol
Inchi Code 1S/C13H11NO4/c1-18-11-5-2-9(3-6-11)12-8-10(14(16)17)4-7-13(12)15/h2-8,15H,1H3
Inchi Key LBOZGNDBSCRERA-UHFFFAOYSA-N

Description

Melting Point 83-84
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11987
Availability details for
Quantity
  • Availability
  • Add to Cart