ENA053246488 Enamine

N-(1,3-dioxaindan-5-yl)-2-chloro-N-{[2-(prop-2-en-1-yloxy)phenyl]methyl}acetamide

  • MDL number MFCD06379302
  • Cas Number : 851452-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11988
Availability details for
Quantity
  • Availability
  • Add to Cart