ENA053246490 Enamine

4,6-dimethylthieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD06660771
  • Cas Number : 885461-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12419

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12419
Availability details for
Quantity
  • Availability
  • Add to Cart