ENA053246492 Enamine

2-chloro-N-(2,3-dihydro-1,4-benzodioxin-6-yl)-N-{[2-(prop-2-en-1-yloxy)phenyl]methyl}acetamide

  • MDL number MFCD06379303
  • Cas Number : 851452-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11989
Availability details for
Quantity
  • Availability
  • Add to Cart