ENA053246497 Enamine

5-[2-chloro-5-(trifluoromethyl)benzyl]-1,3-thiazol-2-amine

  • MDL number MFCD06380308
  • Cas Number : 851398-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11992
Availability details for
Quantity
  • Availability
  • Add to Cart