ENA053246500 Enamine

5-(4-chlorobenzyl)-1,3-thiazol-2-amine

  • MDL number MFCD00696928
  • Cas Number : 290835-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11993

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11993
Availability details for
Quantity
  • Availability
  • Add to Cart