ENA053246501 Enamine

2-[(furan-2-yl)methyl]cyclohexan-1-amine

  • MDL number MFCD00235192
  • Cas Number : 67823-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12430
Availability details for
Quantity
  • Availability
  • Add to Cart