ENA053246504 Enamine

1-(5-chlorofuran-2-yl)methanamine

  • MDL number MFCD06660775
  • Cas Number : 214759-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12433

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-12433
pictograms GHS05,GHS07
hstatements H302,H312,H314
Availability details for
Quantity
  • Availability
  • Add to Cart