ENA053246514 Enamine

[(furan-2-yl)methyl](2,2,2-trifluoroethyl)amine

  • MDL number MFCD06680128
  • Cas Number : 852840-61-0
  • Molecular Weight : 179.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12443
Availability details for
Quantity
  • Availability
  • Add to Cart