ENA053246516 Enamine

4-(1H-imidazol-1-ylmethyl)aniline

  • MDL number MFCD06660785
  • Cas Number : 56643-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12445
Availability details for
Quantity
  • Availability
  • Add to Cart