ENA053246517 Enamine

10,13-dioxa-4-thia-6-azatricyclo[7.4.0.0,3,7]trideca-1,3(7),5,8-tetraen-5-amine

  • MDL number MFCD00560023
  • Cas Number : 313223-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12446
Availability details for
Quantity
  • Availability
  • Add to Cart