ENA053246518 Enamine

5,6-dimethoxy-1,3-benzothiazol-2-amine

  • MDL number MFCD01663342
  • Cas Number : 6294-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12447
Availability details for
Quantity
  • Availability
  • Add to Cart