ENA053246528 Enamine

2,5-dimethyl-1-(5-methyl-1,2-oxazol-3-yl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD06380321
  • Cas Number : 851398-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12007
Availability details for
Quantity
  • Availability
  • Add to Cart