ENA053246532 Enamine

4-(4-methanesulfonylphenyl)-N-(prop-2-en-1-yl)-1,3-thiazol-2-amine

  • MDL number MFCD06380311
  • Cas Number : 851398-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12010
Availability details for
Quantity
  • Availability
  • Add to Cart