ENA053246543 Enamine

(4-chlorophenyl)(phenyl)methylamine

  • MDL number MFCD00044488
  • Cas Number : 28022-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300...305
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12017

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12017
Availability details for
Quantity
  • Availability
  • Add to Cart