ENA053246549 Enamine

2-chloro-N-[1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethyl]acetamide

  • MDL number MFCD06380314
  • Cas Number : 851398-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-12019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12019
Availability details for
Quantity
  • Availability
  • Add to Cart