ENA053246552 Enamine

2-chloro-N-[(4-chlorophenyl)(phenyl)methyl]acetamide

  • MDL number MFCD06380315
  • Cas Number : 92435-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12021
Availability details for
Quantity
  • Availability
  • Add to Cart