ENA053246565 Enamine

3-chloro-N-[(4-ethoxyphenyl)methyl]-4-methoxyaniline

  • MDL number MFCD04595297
  • Cas Number : 851619-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12030
Availability details for
Quantity
  • Availability
  • Add to Cart