ENA053246566 Enamine

3-chloro-4,5-dihydroxybenzaldehyde

  • MDL number MFCD06380366
  • Cas Number : 34098-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234...236
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12031

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12031
Availability details for
Quantity
  • Availability
  • Add to Cart