ENA053246570 Enamine

(2E)-3-(2-phenyl-2H-1,2,3-triazol-4-yl)prop-2-enoic acid

  • MDL number MFCD01104766
  • Cas Number : 6206-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12499
Availability details for
Quantity
  • Availability
  • Add to Cart