ENA053246572 Enamine

N-ethyl-2-(ethylamino)acetamide

  • MDL number MFCD06380313
  • Cas Number : 97454-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12036
Availability details for
Quantity
  • Availability
  • Add to Cart