ENA053246577 Enamine

8-methyl-2-phenylimidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD04971849
  • Cas Number : 850020-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-12041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12041
Availability details for
Quantity
  • Availability
  • Add to Cart