ENA053246586 Enamine

4-chloro-5,6-dimethyl-2-phenylthieno[2,3-d]pyrimidine

  • MDL number MFCD06660832
  • Cas Number : 83548-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12515
Availability details for
Quantity
  • Availability
  • Add to Cart