ENA053246587 Enamine

4-chloro-2,5-diphenylthieno[2,3-d]pyrimidine

  • MDL number MFCD01462067
  • Cas Number : 83548-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12516

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12516
Availability details for
Quantity
  • Availability
  • Add to Cart