ENA053246589 Enamine

1-phenyl-2-sulfanyl-4,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD01848563
  • Cas Number : 2010-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-255
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12518

Safety Information

signalword Danger
pcodes P260,P264,P270,P271,P280,P284,P301+P310,P302+P352,P304+P340,P310,P312,P320,P330,P361,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-12518
s_phrases_eu 36/37/39-45
pictograms GHS06
hstatements H301,H311,H330
Availability details for
Quantity
  • Availability
  • Add to Cart