ENA053246592 Enamine

4-[(furan-2-yl)methyl]-5-(3-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04426475
  • Cas Number : 537017-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12521
Availability details for
Quantity
  • Availability
  • Add to Cart