ENA053246615 Enamine

5-(chloromethyl)-3-[4-(difluoromethoxy)-3-methoxyphenyl]-1,2,4-oxadiazole

  • MDL number MFCD06655336
  • Cas Number : 852399-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12544
Availability details for
Quantity
  • Availability
  • Add to Cart