ENA053246618 Enamine

2-[5-(4-fluorophenyl)-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-3-yl]acetic acid

  • MDL number MFCD03016018
  • Cas Number : 451461-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12547
Availability details for
Quantity
  • Availability
  • Add to Cart